Assay Details
| Gene Symbol: UQCRB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185172.1 | NP_001172101.1 | 2 - 3 | 121 | 83 | |
| XM_021088627.1 | XP_020944286.1 | 168 | 83 | |||
| GenBank mRNA | AK344783.1 | - | 2 - 3 | 97 | 83 | |
| AY610483.1 | - | 1 - 2 | 97 | 83 | ||
| EV895402.2 | - | 1 - 2 | 48 | 83 | ||
| EV897222.2 | - | 2 - 3 | 130 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021088627.1 transcripts.
Set Membership: |
|

UQCRB
Genomic Map