Assay Details
| Gene Symbol: RGS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185184.1 | NP_001172113.1 | 1 - 2 | 165 | 63 | |
| XM_021088597.1 | XP_020944256.1 | 179 | 63 | |||
| GenBank mRNA | AK231087.1 | - | 1 - 2 | 165 | 63 | |
| AK391737.1 | - | 1 - 2 | 167 | 63 | ||
More Information
Set Membership: |
|

RGS4
Genomic Map