Assay Details
| Gene Symbol: TMEM38B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190164.1 | NP_001177093.1 | 1 - 2 | 165 | 61 | |
| GenBank mRNA | AY609714.1 | - | 1 - 2 | 165 | 61 | |
More Information
Set Membership: |
|

TMEM38B
Genomic Map