Assay Details
| Gene Symbol: CDK2AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190166.1 | NP_001177095.1 | 2 - 3 | 409 | 94 | |
| XM_021071919.1 | XP_020927578.1 | 209 | 94 | |||
| GenBank mRNA | AK232148.1 | - | 2 - 3 | 409 | 94 | |
| AK345389.1 | - | 2 - 3 | 409 | 94 | ||
More Information
Set Membership: |
|

CDK2AP1
Genomic Map