Assay Details
| Gene Symbol: DIABLO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190216.1 | NP_001177145.1 | 1 - 2 | 103 | 59 | |
| GenBank mRNA | AK234061.1 | - | 1 - 2 | 87 | 59 | |
| AK346394.1 | - | 1 - 2 | 87 | 59 | ||
| AK389815.1 | - | 1 - 2 | 88 | 59 | ||
| AY609440.1 | - | 1 - 2 | 64 | 59 | ||
| DT330216.1 | - | 1 - 2 | 122 | 59 | ||
| FJ234415.1 | - | 1 - 2 | 56 | 59 | ||
More Information
Set Membership: |
|

DIABLO
Genomic Map