Assay Details
| Gene Symbol: USF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190245.1 | NP_001177174.1 | 9 - 10 | 893 | 77 | |
| XM_005663180.3 | XP_005663237.1 | 959 | 77 | |||
| XM_005663182.3 | XP_005663239.1 | 948 | 77 | |||
| XM_013996843.2 | XP_013852297.1 | 924 | 77 | |||
| XM_013996844.2 | XP_013852298.1 | 970 | 77 | |||
| XM_013996845.2 | XP_013852299.1 | 772 | 77 | |||
| XM_021088586.1 | XP_020944245.1 | 1198 | 77 | |||
| XM_021088587.1 | XP_020944246.1 | 1085 | 77 | |||
| XM_021088588.1 | XP_020944247.1 | 957 | 77 | |||
| XM_021088589.1 | XP_020944248.1 | 1007 | 77 | |||
| XM_021088590.1 | XP_020944249.1 | 840 | 77 | |||
| XM_021088591.1 | XP_020944250.1 | 975 | 77 | |||
| XM_021088592.1 | XP_020944251.1 | 1099 | 77 | |||
| XM_021088593.1 | XP_020944252.1 | 854 | 77 | |||
| GenBank mRNA | AK398370.1 | - | 9 - 10 | 892 | 77 | |
| BP438739.1 | - | 1 - 2 | 100 | 77 | ||
| EF208923.1 | - | 8 - 9 | 601 | 77 | ||
| EF219407.1 | - | 8 - 9 | 784 | 77 | ||
More Information
Set Membership: |
|

USF1
Genomic Map