Assay Details
| Gene Symbol: WIPI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190294.1 | NP_001177223.1 | 9 - 10 | 1015 | 58 | |
| GenBank mRNA | AK233423.1 | - | 9 - 10 | 1015 | 58 | |
| AK396744.1 | - | 9 - 10 | 1029 | 58 | ||
| AY609688.1 | - | 9 - 10 | 948 | 58 | ||
| HM046516.1 | - | 9 - 10 | 1015 | 58 | ||
More Information
Set Membership: |
|

WIPI1
Genomic Map