Assay Details
| Gene Symbol: GPR33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190923.1 | NP_001177852.1 | 1 - 2 | 137 | 82 | |
| GenBank mRNA | GQ981325.1 | - | 1 - 2 | 137 | 82 | |
| GQ981326.1 | - | 1 - 2 | 137 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001190923.1 transcripts.
Set Membership: |
|

GPR33
Genomic Map