Assay Details
| Gene Symbol: C12H17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195120.1 | NP_001182049.1 | 2 - 3 | 262 | 85 | |
| XM_021066070.1 | XP_020921729.1 | 316 | 85 | |||
| XM_021066071.1 | XP_020921730.1 | 308 | 85 | |||
| GenBank mRNA | HM627269.1 | - | 2 - 3 | 262 | 85 | |
More Information
Set Membership: |
|

C12H17orf75
Genomic Map