Assay Details
| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204181.2 | NP_001191110.1 | 2 - 3 | 2552 | 69 | |
| GenBank mRNA | AK348182.1 | - | 2 - 3 | 2551 | 69 | |
More Information
Set Membership: |
|

PALM2AKAP2
Genomic Map