Assay Details
| Gene Symbol: PALM2AKAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204195.2 | NP_001191124.2 | 6 - 7 | 755 | 98 | |
| NM_001370837.1 | NP_001357766.1 | 6 - 7 | 755 | 98 | ||
| GenBank mRNA | AK234430.1 | - | 6 - 7 | 784 | 98 | |
| AK347588.1 | - | 6 - 7 | 755 | 98 | ||
More Information
Set Membership: |
|

PALM2AKAP2
Genomic Map