Assay Details
| Gene Symbol: PPAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204435.1 | NP_001191364.1 | 5 - 6 | 577 | 63 | |
| GenBank mRNA | BP168379.1 | - | 5 - 6 | 542 | 63 | |
More Information
Set Membership: |
|

PPAN
Genomic Map