Assay Details
| Gene Symbol: FFAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204766.2 | NP_001191695.1 | 1 - 2 | 619 | 77 | |
| Gene Symbol: PDE6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | HQ662564.1 | - | 1 - 2 | 578 | 77 | |
| JQ437359.1 | - | 1 - 2 | 588 | 77 | ||
More Information
Set Membership: |
|

FFAR4
Genomic Map