Assay Details
| Gene Symbol: FFAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204766.2 | NP_001191695.1 | 2 - 3 | 740 | 108 | |
| Gene Symbol: PDE6C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | HQ662564.1 | - | 2 - 3 | 699 | 108 | |
| JQ437359.1 | - | 2 - 3 | 709 | 108 | ||
| JQ437361.1 | - | 2 - 3 | 553 | 108 | ||
More Information
Set Membership: |
|

FFAR4
Genomic Map