Assay Details
| Gene Symbol: APITD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204935.1 | NP_001191864.1 | 1 - 2 | 87 | 60 | |
| XM_005664980.3 | XP_005665037.1 | 172 | 60 | |||
| XM_021093344.1 | XP_020949003.1 | 979 | 60 | |||
| GenBank mRNA | AK351775.1 | - | 1 - 2 | 104 | 60 | |
| CK458744.1 | - | 1 - 2 | 87 | 60 | ||
More Information
Set Membership: |
|

APITD1
Genomic Map