Assay Details
| Gene Symbol: APITD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204935.1 | NP_001191864.1 | 4 - 5 | 308 | 102 | |
| GenBank mRNA | CK458744.1 | - | 4 - 5 | 308 | 102 | |
More Information
Set Membership: |
|

APITD1
Genomic Map