Assay Details
| Gene Symbol: H1FOO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205063.1 | NP_001191992.1 | 2 - 3 | 428 | 79 | |
| XM_021068419.1 | XP_020924078.1 | 417 | 79 | |||
| GenBank mRNA | HQ915640.1 | - | 2 - 3 | 428 | 79 | |
More Information
Set Membership: |
|

H1FOO
Genomic Map