Assay Details
| Gene Symbol: ZDHHC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003122494.6 | XP_003122542.3 | 396 | 54 | ||
| GenBank mRNA | AK235222.1 | - | 1 - 2 | 346 | 54 | |
| AK240521.1 | - | 1 - 2 | 398 | 54 | ||
More Information
Set Membership: |
|

ZDHHC24
Genomic Map