Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163408.1 | NP_001156880.1 | 1 - 2 | 176 | 58 | |
| GenBank mRNA | AK237421.1 | - | 1 - 2 | 239 | 58 | |
| AK399219.1 | - | 1 - 2 | 231 | 58 | ||
| AY609428.2 | - | 1 - 2 | 221 | 58 | ||
| GQ140624.1 | - | 1 - 2 | 176 | 58 | ||
More Information
Set Membership: |
|

SELENOT
Genomic Map