Assay Details
| Gene Symbol: SELENOT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163408.1 | NP_001156880.1 | 2 - 3 | 272 | 73 | |
| GenBank mRNA | AK237421.1 | - | 2 - 3 | 335 | 73 | |
| AK399219.1 | - | 2 - 3 | 327 | 73 | ||
| AY609428.2 | - | 2 - 3 | 317 | 73 | ||
| GQ140624.1 | - | 2 - 3 | 272 | 73 | ||
More Information
Set Membership: |
|

SELENOT
Genomic Map