Assay Details
| Gene Symbol: UQCC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172364.2 | NP_001165835.1 | 2 - 3 | 150 | 58 | |
| XM_021077328.1 | XP_020932987.1 | 262 | 58 | |||
| GenBank mRNA | AK236572.1 | - | 2 - 3 | 147 | 58 | |
| AK343765.1 | - | 2 - 3 | 141 | 58 | ||
| AK391617.1 | - | 2 - 3 | 237 | 58 | ||
| GU373690.1 | - | 2 - 3 | 151 | 58 | ||
More Information
Set Membership: |
|

UQCC1
Genomic Map