Assay Details
| Gene Symbol: SOCS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204768.1 | NP_001191697.1 | 1 | 724 | 65 | |
| XM_005662125.3 | XP_005662182.1 | 880 | 65 | |||
| XM_005662126.3 | XP_005662183.1 | 2374 | 65 | |||
| GenBank mRNA | GQ421919.1 | - | 1 | 473 | 65 | |
| HM462248.1 | - | 1 | 724 | 65 | ||
More Information
Set Membership: |
|

SOCS1
Genomic Map