Assay Details
| Gene Symbol: POLR2I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205404.1 | NP_001192333.1 | 3 - 4 | 556 | 65 | |
| NM_001205406.1 | NP_001192335.1 | 2 - 3 | 277 | 65 | ||
| GenBank mRNA | AK234376.1 | - | 3 - 4 | 218 | 65 | |
| AK400001.1 | - | 2 - 3 | 277 | 65 | ||
| EW246615.2 | - | 2 - 3 | 288 | 65 | ||
| EW379062.2 | - | 2 - 3 | 296 | 65 | ||
| FD595099.1 | - | 3 - 4 | 556 | 65 | ||
More Information
Set Membership: |
|

POLR2I
Genomic Map