Assay Details
| Gene Symbol: ATP5MC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244862.1 | NP_001231791.1 | 3 - 4 | 245 | 106 | |
| GenBank mRNA | AK232718.1 | - | 3 - 4 | 263 | 106 | |
| AK348215.1 | - | 3 - 4 | 241 | 106 | ||
| GQ913653.1 | - | 2 - 3 | 187 | 106 | ||
More Information
Set Membership: |
|

ATP5MC3
Genomic Map