Assay Details
| Gene Symbol: SDCBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244863.1 | NP_001231792.1 | 4 - 5 | 385 | 58 | |
| XM_013985196.2 | XP_013840650.1 | 717 | 58 | |||
| XM_021077266.1 | XP_020932925.1 | 1382 | 58 | |||
| XM_021077267.1 | XP_020932926.1 | 662 | 58 | |||
| GenBank mRNA | AK349737.1 | - | 4 - 5 | 387 | 58 | |
More Information
Set Membership: |
|

SDCBP2
Genomic Map