Assay Details
| Gene Symbol: NDUFB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244885.1 | NP_001231814.1 | 2 - 3 | 253 | 77 | |
| GenBank mRNA | AK392487.1 | - | 2 - 3 | 284 | 77 | |
| AK399891.1 | - | 2 - 3 | 292 | 77 | ||
| AY610262.1 | - | 2 - 3 | 253 | 77 | ||
More Information
Set Membership: |
|

NDUFB2
Genomic Map