Assay Details
| Gene Symbol: LRAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244920.1 | NP_001231849.1 | 2 - 3 | 775 | 90 | |
| XM_005666789.3 | XP_005666846.1 | 793 | 90 | |||
| XM_005666790.3 | XP_005666847.1 | 616 | 90 | |||
| XM_021100353.1 | XP_020956012.1 | 924 | 90 | |||
| GenBank mRNA | AK239291.1 | - | 2 - 3 | 775 | 90 | |
More Information
Set Membership: |
|

LRAT
Genomic Map