Assay Details
| Gene Symbol: NDUFA11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001244924.1 | NP_001231853.1 | 3 - 4 | 387 | 107 | |
| GenBank mRNA | AK351136.1 | - | 3 - 4 | 1013 | 107 | |
| AK391721.1 | - | 3 - 4 | 361 | 107 | ||
| FD598063.1 | - | 3 - 4 | 387 | 107 | ||
More Information
Set Membership: |
|

NDUFA11
Genomic Map