Assay Details
| Gene Symbol: MRPL49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001245013.1 | NP_001231942.1 | 1 - 2 | 103 | 102 | |
| XM_021078948.1 | XP_020934607.1 | 752 | 102 | |||
| GenBank mRNA | AK236704.1 | - | 1 - 2 | 103 | 102 | |
More Information
Set Membership: |
|

MRPL49
Genomic Map