Assay Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246198.2 | NP_001233127.2 | 10 - 11 | 1036 | 98 | |
| GenBank mRNA | AK237133.1 | - | 9 - 10 | 1007 | 98 | |
| AK347838.1 | - | 10 - 11 | 885 | 98 | ||
| AK398121.1 | - | 10 - 11 | 1035 | 98 | ||
| DB816484.1 | - | 3 - 4 | 322 | 98 | ||
| EU650400.1 | - | 10 - 11 | 876 | 98 | ||
More Information
Set Membership: |
|

TDRD10
Genomic Map