Assay Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246198.2 | NP_001233127.2 | 11 - 12 | 1193 | 81 | |
| GenBank mRNA | AK237133.1 | - | 10 - 11 | 1164 | 81 | |
| AK347838.1 | - | 11 - 12 | 1042 | 81 | ||
| AK398121.1 | - | 11 - 12 | 1192 | 81 | ||
| DB816484.1 | - | 2 - 3 | 165 | 81 | ||
| DY406885.1 | - | 2 - 3 | 469 | 81 | ||
| EU650400.1 | - | 11 - 12 | 81 | |||
| JZ566346.1 | - | 1 - 2 | 167 | 81 | ||
More Information
Set Membership: |
|

TDRD10
Genomic Map