Assay Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246198.2 | NP_001233127.2 | 12 - 13 | 1266 | 73 | |
| GenBank mRNA | AK237133.1 | - | 11 - 12 | 1237 | 73 | |
| AK347838.1 | - | 12 - 13 | 1115 | 73 | ||
| AK398121.1 | - | 12 - 13 | 1265 | 73 | ||
| DB816484.1 | - | 1 - 2 | 92 | 73 | ||
| DY406885.1 | - | 1 - 2 | 396 | 73 | ||
| EU650400.1 | - | 12 - 13 | 1106 | 73 | ||
| JZ566346.1 | - | 2 - 3 | 240 | 73 | ||
More Information
Set Membership: |
|

TDRD10
Genomic Map