Assay Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246198.2 | NP_001233127.2 | 3 - 4 | 317 | 90 | |
| GenBank mRNA | AK237133.1 | - | 2 - 3 | 288 | 90 | |
| AK347838.1 | - | 3 - 4 | 166 | 90 | ||
| AK398121.1 | - | 3 - 4 | 316 | 90 | ||
| EU650400.1 | - | 3 - 4 | 157 | 90 | ||
| FD640253.1 | - | 3 - 4 | 90 | |||
More Information
Set Membership: |
|

TDRD10
Genomic Map