Assay Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246198.2 | NP_001233127.2 | 4 - 5 | 376 | 83 | |
| GenBank mRNA | AK237133.1 | - | 3 - 4 | 347 | 83 | |
| AK347838.1 | - | 4 - 5 | 225 | 83 | ||
| AK398121.1 | - | 4 - 5 | 375 | 83 | ||
| EU650400.1 | - | 4 - 5 | 216 | 83 | ||
| FD640253.1 | - | 4 - 5 | 376 | 83 | ||
More Information
Set Membership: |
|

TDRD10
Genomic Map