Assay Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246198.2 | NP_001233127.2 | 7 - 8 | 651 | 62 | |
| GenBank mRNA | AK237133.1 | - | 6 - 7 | 622 | 62 | |
| AK347838.1 | - | 7 - 8 | 500 | 62 | ||
| AK398121.1 | - | 7 - 8 | 650 | 62 | ||
| DB816484.1 | - | 6 - 7 | 707 | 62 | ||
| EU650400.1 | - | 7 - 8 | 491 | 62 | ||
More Information
Set Membership: |
|

TDRD10
Genomic Map