Assay Details
| Gene Symbol: TDRD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246198.2 | NP_001233127.2 | 9 - 10 | 887 | 83 | |
| GenBank mRNA | AK237133.1 | - | 8 - 9 | 858 | 83 | |
| AK347838.1 | - | 9 - 10 | 736 | 83 | ||
| AK398121.1 | - | 9 - 10 | 886 | 83 | ||
| DB816484.1 | - | 4 - 5 | 471 | 83 | ||
| EU650400.1 | - | 9 - 10 | 727 | 83 | ||
More Information
Set Membership: |
|

TDRD10
Genomic Map