Assay Details
| Gene Symbol: ANKS1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001246264.2 | NP_001233193.1 | 11 - 12 | 1483 | 64 | |
| XM_021098214.1 | XP_020953873.1 | 1619 | 64 | |||
| XM_021098215.1 | XP_020953874.1 | 1619 | 64 | |||
| XM_021098216.1 | XP_020953875.1 | 1555 | 64 | |||
| XM_021098217.1 | XP_020953876.1 | 1526 | 64 | |||
| XM_021098218.1 | XP_020953877.1 | 1618 | 64 | |||
| XM_021098219.1 | XP_020953878.1 | 1618 | 64 | |||
| XM_021098220.1 | XP_020953879.1 | 1617 | 64 | |||
| XM_021098221.1 | XP_020953880.1 | 1616 | 64 | |||
| XM_021098222.1 | XP_020953881.1 | 1553 | 64 | |||
| XM_021098223.1 | XP_020953882.1 | 1586 | 64 | |||
| XM_021098224.1 | XP_020953883.1 | 3495 | 64 | |||
| GenBank mRNA | HQ630074.1 | - | 11 - 12 | 1483 | 64 | |
More Information
Set Membership: |
|

ANKS1A
Genomic Map