Assay Details
| Gene Symbol: RAB10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243060.1 | NP_001229989.1 | 5 - 6 | 1051 | 83 | |
| GenBank mRNA | AK235234.1 | - | 7 - 8 | 982 | 83 | |
| AK237668.1 | - | 5 - 6 | 1051 | 83 | ||
| AK237790.1 | - | 5 - 6 | 1043 | 83 | ||
| AK350491.1 | - | 5 - 6 | 981 | 83 | ||
| AK395567.1 | - | 5 - 6 | 1058 | 83 | ||
| AY609790.1 | - | 5 - 6 | 875 | 83 | ||
More Information
Set Membership: |
|

RAB10
Genomic Map