Assay Details
| Gene Symbol: ARV1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243293.1 | NP_001230222.1 | 5 - 6 | 880 | 70 | |
| GenBank mRNA | AK348219.1 | - | 5 - 6 | 880 | 70 | |
| AK391940.1 | - | 5 - 6 | 865 | 70 | ||
| AY610027.1 | - | 5 - 6 | 825 | 70 | ||
| FS700419.1 | - | 5 - 6 | 846 | 70 | ||
More Information
Set Membership: |
|

ARV1
Genomic Map