Assay Details
| Gene Symbol: FRZB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243330.1 | NP_001230259.1 | 5 - 6 | 1021 | 63 | |
| XM_021074512.1 | XP_020930171.1 | 1272 | 63 | |||
| GenBank mRNA | AK236391.1 | - | 5 - 6 | 1085 | 63 | |
| AY609868.1 | - | 5 - 6 | 923 | 63 | ||
| KF040089.1 | - | 5 - 6 | 1648 | 63 | ||
More Information
Set Membership: |
|

FRZB
Genomic Map