Assay Details
| Gene Symbol: SHOX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243352.1 | NP_001230281.1 | 1 - 2 | 352 | 90 | |
| XM_005654033.3 | XP_005654090.1 | 3042 | 90 | |||
More Information
Set Membership: |
|

SHOX2
Genomic Map