Assay Details
| Gene Symbol: METTL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243367.1 | NP_001230296.1 | 2 - 3 | 542 | 96 | |
| XM_021085766.1 | XP_020941425.1 | 611 | 96 | |||
| XM_021085767.1 | XP_020941426.1 | 334 | 96 | |||
| XM_021085768.1 | XP_020941427.1 | 334 | 96 | |||
| GenBank mRNA | AK350557.1 | - | 2 - 3 | 542 | 96 | |
More Information
Set Membership: |
|

METTL9
Genomic Map