Assay Details
| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243371.1 | NP_001230300.1 | 3 - 4 | 382 | 95 | |
| GenBank mRNA | AK347930.1 | - | 3 - 4 | 432 | 95 | |
| CF177255.1 | - | 3 - 4 | 382 | 95 | ||
| FS645850.1 | - | 1 - 2 | 467 | 95 | ||
More Information
Set Membership: |
|

VPS37B
Genomic Map