Assay Details
| Gene Symbol: LIN37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243377.1 | NP_001230306.1 | 5 - 6 | 422 | 88 | |
| XM_013998326.2 | XP_013853780.1 | 538 | 88 | |||
| XM_013998328.2 | XP_013853782.1 | 545 | 88 | |||
| XM_021093436.1 | XP_020949095.1 | 459 | 88 | |||
| GenBank mRNA | AK231288.1 | - | 5 - 6 | 422 | 88 | |
| AK231399.1 | - | 5 - 6 | 422 | 88 | ||
| AK352403.1 | - | 5 - 6 | 422 | 88 | ||
More Information
Set Membership: |
|

LIN37
Genomic Map