Assay Details
| Gene Symbol: PPP1R8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243414.1 | NP_001230343.1 | 6 - 7 | 768 | 79 | |
| XM_021093434.1 | XP_020949093.1 | 513 | 79 | |||
| GenBank mRNA | BP464075.1 | - | 6 - 7 | 728 | 79 | |
| EW578770.2 | - | 2 - 3 | 186 | 79 | ||
More Information
Set Membership: |
|

PPP1R8
Genomic Map