Assay Details
| Gene Symbol: JOSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243416.1 | NP_001230345.1 | 2 - 3 | 320 | 75 | |
| XM_013998690.2 | XP_013854144.1 | 343 | 75 | |||
| XM_021093433.1 | XP_020949092.1 | 344 | 75 | |||
| GenBank mRNA | AK236047.1 | - | 2 - 3 | 326 | 75 | |
| AK396597.1 | - | 2 - 3 | 239 | 75 | ||
| AK400224.1 | - | 2 - 3 | 348 | 75 | ||
| DT319721.1 | - | 2 - 3 | 320 | 75 | ||
More Information
Set Membership: |
|

JOSD2
Genomic Map