Assay Details
| Gene Symbol: HAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243431.1 | NP_001230360.1 | 1 - 2 | 179 | 63 | |
| XM_021077275.1 | XP_020932934.1 | 175 | 63 | |||
| GenBank mRNA | AK232269.1 | - | 1 - 2 | 163 | 63 | |
| AK393986.1 | - | 1 - 2 | 172 | 63 | ||
| FD604698.1 | - | 1 - 2 | 181 | 63 | ||
More Information
Set Membership: |
|

HAO1
Genomic Map