Assay Details
| Gene Symbol: TMIGD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243436.2 | NP_001230365.1 | 5 - 6 | 691 | 65 | |
| GenBank mRNA | CV869126.1 | - | 4 - 5 | 629 | 65 | |
| CX056678.1 | - | 2 - 3 | 186 | 65 | ||
More Information
Set Membership: |
|

TMIGD3
Genomic Map