Assay Details
| Gene Symbol: AP4S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243497.1 | NP_001230426.1 | 2 - 3 | 303 | 101 | |
| XM_021098033.1 | XP_020953692.1 | 4435 | 101 | |||
| XM_021098034.1 | XP_020953693.1 | 4435 | 101 | |||
| XM_021098036.1 | XP_020953695.1 | 4435 | 101 | |||
| XM_021098037.1 | XP_020953696.1 | 4435 | 101 | |||
| GenBank mRNA | AK345147.1 | - | 3 - 4 | 438 | 101 | |
More Information
Set Membership: |
|

AP4S1
Genomic Map