Assay Details
Gene Symbol: SMIM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243508.1 | NP_001230437.1 | 2 - 3 | 211 | 76 | |
XM_005659276.3 | XP_005659333.1 | 255 | 76 | |||
GenBank mRNA | AK347103.1 | - | 2 - 3 | 211 | 76 | |
AK399623.1 | - | 2 - 3 | 211 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
